Learn More
Aviva Systems Biology Corp ATOH7 Antibody - C-terminal region (ARP36152_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36152_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATOH7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ATOH7. Alternate names: Math5, bHLHa13ApplicationWB. Species reactivity: Cow, Dog, Human, Pig
Catalog No. 50-014-56034