Learn More
Aviva Systems Biology Corp ATOH8 Antibody - C-terminal region (ARP39729_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39729_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATOH8. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ATOH8. Alternate names: FLJ14708, FLJ38730, HATH6, bHLHa21ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-824