Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Atomic Model Set
GREENER_CHOICESCI_ED

Catalog No. S139416 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139416 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139416 Supplier Supertek™ Supplier No. CH11560/4
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Supertek™ Atomic Model Set consists of molded balls in different colors and sizes along with connectors of different sizes. The sizes and shapes of the balls and connecting lugs have been designed in accordance with internationally acceptable standards.

  • 75 large atoms in 6 colors.
  • 25 small atoms in 3 colors.
  • 36 large connectors
  • 51 small connectors
  • 7 constructed models denoted on a teacher reference chart.
  • Handy plastic storage container.

Specifications

Grade 6+
Format Class Set
Languages English
Subject Chemistry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.