Learn More
Electron Microscopy Sciences Sure Shot Atomizer Model 8000CB Plated Red Powder Coated Aluminum, Chrome-Plated Brass Top 16 OZ
SDP
Catalog No. 501928729
Encompass_Preferred
Anything that the aerosol can do, ATOMIZER can do safer. Refillable and Rechargeable. Eliminates aerosol disposal problems. All Model B sprayers come standard with Viton seals, these seals are compatible with most products. IMPORTANT: Sure Shot Sprayers are not recommended for spraying any acids. Sure Shot Sprayers will handle most chemicals, but are not designed to be used with products that are intended to etch or attack metal surfaces. Plated Red Powder coated aluminum and chrome-plated brass top. Comes with extra valve and 2 fine, 2 regular, 2 medium, 2 coarse and 1 pin stream nozzle.
Catalog No. 50-192-8729 Supplier Electron Microscopy Sciences Supplier No. 6418604
Anything that the aerosol can do, ATOMIZER can do safer. Refillable and Rechargeable. Eliminates aerosol disposal problems. All Model B sprayers come standard with Viton seals, these seals are compatible with most products. IMPORTANT: Sure Shot Sprayers are not recommended for spraying any acids. Sure Shot Sprayers will handle most chemicals, but are not designed to be used with products that are intended to etch or attack metal surfaces. Plated Red Powder coated aluminum and chrome-plated brass top. Comes with extra valve and 2 fine, 2 regular, 2 medium, 2 coarse and 1 pin stream nozzle.