Learn More
Electron Microscopy Sciences Atomizer Rubber Bulb, 45cc
SDP
Catalog No. 50996279
Encompass_Preferred
A natural rubber bulb with 4" tubing on one end. Used for dusting surfaces which should not be wiped. A two-way valve is inserted into one end to prevent suck back and a rubber nozzle prevents scratches on the surface that have to be cleaned. This device also reduces the need for freon dusters.
Catalog No. 50-996-279 Supplier Electron Microscopy Sciences Supplier No. 60804
A natural rubber bulb with 4" tubing on one end. Used for dusting surfaces which should not be wiped. A two-way valve is inserted into one end to prevent suck back and a rubber nozzle prevents scratches on the surface that have to be cleaned. This device also reduces the need for freon dusters.