Learn More
TARGETMOL CHEMICALS INC Atomoxetine hydrochloride
SDP
Catalog No. 502880707
Encompass_Preferred
Also available in 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Atomoxetine hydrochloride (LY 139603) is the hydrochloride salt of atomoxetine, a phenoxy-3-propylamine derivative and selective non-stimulant, norepinephrine reuptake inhibitor with cognitive-enhancing activity. Although its precise mechanism of action is unknown, atomoxetine appears to selectively inhibit the pre-synaptic norepinephrine transporter, resulting in inhibition of the presynaptic reabsorption of norepinephrine and prolongation of norepinephrine activity in the synaptic cleft. The effect on cognitive brain function may result in improved attention and decreased impulsivity and activity levels. Purity 99.8%
Catalog No. 50-288-0707 Supplier TARGETMOL CHEMICALS INC Supplier No. T0869200MG
Also available in 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Atomoxetine hydrochloride (LY 139603) is the hydrochloride salt of atomoxetine, a phenoxy-3-propylamine derivative and selective non-stimulant, norepinephrine reuptake inhibitor with cognitive-enhancing activity. Although its precise mechanism of action is unknown, atomoxetine appears to selectively inhibit the pre-synaptic norepinephrine transporter, resulting in inhibition of the presynaptic reabsorption of norepinephrine and prolongation of norepinephrine activity in the synaptic cleft. The effect on cognitive brain function may result in improved attention and decreased impulsivity and activity levels. Purity 99.8%