Learn More
Medchemexpress LLC Atomoxetine hydrochloride | 82248-59-7 | 99.9% | 291.82 | 500 MG
SDP
Catalog No. 5000384779
Encompass_Preferred
Atomoxetine hydrochloride is a selective noradrenaline reuptake inhibitor that can be used for attention-deficit hyperactivity disorder (ADHD) research. It acts as a potent Na+ channels (VGSCs) blocker and interacts with the human heart muscle sodium channel (hNav1.5).
  • Selective noradrenaline reuptake inhibitor
  • Potent Na+ channels (VGSCs) blocker
  • Suitable for ADHD research
  • Interacts with human heart muscle sodium channel
Catalog No. 50-003-84779 Supplier Medchemexpress LLC Supplier No. HY17385500MG
Atomoxetine hydrochloride is a selective noradrenaline reuptake inhibitor that can be used for attention-deficit hyperactivity disorder (ADHD) research. It acts as a potent Na+ channels (VGSCs) blocker and interacts with the human heart muscle sodium channel (hNav1.5).
  • Selective noradrenaline reuptake inhibitor
  • Potent Na+ channels (VGSCs) blocker
  • Suitable for ADHD research
  • Interacts with human heart muscle sodium channel