Learn More
Medchemexpress LLC Atopaxar | 751475-53-3 | 99.9% | 527.63 | 5 MG
SDP
Catalog No. 5000387481
Encompass_Preferred
Atopaxar is a potent, orally active, selective and reversible thrombin receptor protease-activated receptor-1 (PAR-1) antagonist. As an antiplatelet agent, it interferes with platelet signaling and can be used for the research of atherothrombotic disease.
  • Potent, orally active, selective and reversible PAR-1 antagonist
  • Interferes with platelet signaling
  • Used for research of atherothrombotic disease
  • Inhibits human platelet aggregation induced by thrombin or TRAP
  • Does not inhibit PRP aggregation induced by ADP, U46619, collagen, and PAR-4ap, up to 20 μM
  • Causes dose-dependent prolongation of time to occlusion of the femoral artery in photochemically-induced thrombosis (PIT) guinea pigs model
  • Does not prolong bleeding time in guinea pigs at high dosages
Catalog No. 50-003-87481 Supplier Medchemexpress LLC Supplier No. HY182005MG
Atopaxar is a potent, orally active, selective and reversible thrombin receptor protease-activated receptor-1 (PAR-1) antagonist. As an antiplatelet agent, it interferes with platelet signaling and can be used for the research of atherothrombotic disease.
  • Potent, orally active, selective and reversible PAR-1 antagonist
  • Interferes with platelet signaling
  • Used for research of atherothrombotic disease
  • Inhibits human platelet aggregation induced by thrombin or TRAP
  • Does not inhibit PRP aggregation induced by ADP, U46619, collagen, and PAR-4ap, up to 20 μM
  • Causes dose-dependent prolongation of time to occlusion of the femoral artery in photochemically-induced thrombosis (PIT) guinea pigs model
  • Does not prolong bleeding time in guinea pigs at high dosages