Learn More
Medchemexpress LLC Atorvastatin | 134523-00-5 | 99.60% | 558.64 | 100 MG
SDP
Catalog No. 5000364748
Encompass_Preferred
Atorvastatin is an orally active HMG-CoA reductase inhibitor that effectively decreases blood lipids. It inhibits human SV-SMC proliferation and invasion, and decreases apoptosis of myocardial cells by down-regulating GRP78, caspase-12, and CHOP expression after myocardial infarction. It also activates endoplasmic reticulum (ER) stress in response to heart failure and angiotensin II (Ang II) stimulation.
  • Orally active HMG-CoA reductase inhibitor
  • Effectively decreases blood lipids
  • Inhibits human SV-SMC proliferation and invasion
  • Decreases apoptosis of myocardial cells
  • Down-regulates GRP78, caspase-12, and CHOP expression
  • Activates endoplasmic reticulum (ER) stress
Catalog No. 50-003-64748 Supplier Medchemexpress LLC Supplier No. 100MGHYB0589
Atorvastatin is an orally active HMG-CoA reductase inhibitor that effectively decreases blood lipids. It inhibits human SV-SMC proliferation and invasion, and decreases apoptosis of myocardial cells by down-regulating GRP78, caspase-12, and CHOP expression after myocardial infarction. It also activates endoplasmic reticulum (ER) stress in response to heart failure and angiotensin II (Ang II) stimulation.
  • Orally active HMG-CoA reductase inhibitor
  • Effectively decreases blood lipids
  • Inhibits human SV-SMC proliferation and invasion
  • Decreases apoptosis of myocardial cells
  • Down-regulates GRP78, caspase-12, and CHOP expression
  • Activates endoplasmic reticulum (ER) stress