Learn More
Medchemexpress LLC Atorvastatin | 134523-00-5 | MFCD00868840 | >98% | 558.64 | 10 MG
SDP
Supplier:  Medchemexpress LLC 10MGHYB0589
Encompass_Preferred
Atorvastatin is an orally active HMG-CoA reductase inhibitor that effectively decreases blood lipids. It inhibits human SV-SMC proliferation and invasion with IC50s of 0.39 μM and 2.39 μM, respectively. Atorvastatin also possesses potential anti-inflammatory, antioxidant, and anti-proliferative effects.
  • Effectively decreases blood lipids.
  • Inhibits human SV-SMC proliferation and invasion.
  • Potential anti-inflammatory, antioxidant, and anti-proliferative effects.
  • Target: HMG-CoA Reductase (HMGCR), Autophagy.
  • Research Areas: Metabolic Disease, Inflammation/Immunology, Cancer.
Catalog No. 50-003-65412