Learn More
Medchemexpress LLC Atorvastatin | 134523-00-5 | MFCD00899261 | 99.6% | 500 MG
SDP
Catalog No. 5000408353
Encompass_Preferred
Atorvastatin is an orally active HMG-CoA reductase inhibitor that effectively decreases blood lipids. It inhibits human SV-SMC proliferation and invasion with IC50s of 0.39 μM and 2.39 μM, respectively. It is intended for research use only and is not sold to patients.
  • Orally active HMG-CoA reductase inhibitor
  • Effectively decreases blood lipids
  • Inhibits human SV-SMC proliferation and invasion
Catalog No. 50-004-08353 Supplier Medchemexpress LLC Supplier No. HYB0589500MG
Atorvastatin is an orally active HMG-CoA reductase inhibitor that effectively decreases blood lipids. It inhibits human SV-SMC proliferation and invasion with IC50s of 0.39 μM and 2.39 μM, respectively. It is intended for research use only and is not sold to patients.
  • Orally active HMG-CoA reductase inhibitor
  • Effectively decreases blood lipids
  • Inhibits human SV-SMC proliferation and invasion