Learn More
Selleck Chemical LLC atorvastatin 25mg 110862-48-1
SDP
Supplier:  Selleck Chemical LLC S571525MG
Encompass_Preferred
Atorvastatin is a lipid lowering agent. It is a competitive inhibitor of hydroxymethylglutaryl-coenzyme A (HMG-CoA) reductase, the rate-determining enzyme in cholesterol biosynthesis via the mevalonate pathway. Atorvastatin activates autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8178