Learn More
Medchemexpress LLC HY-B0589 50mg , Atorvastatin CAS:134523-00-5 Purity:>98%
SDP
Catalog No. 502259576
Encompass_Preferred
HY-B0589 50mg Atorvastatin CAS: 134523-00-5 Atorvastatin is an orally active HMG-CoA reductase inhibitor, has the ability to effectively decrease blood lipids. Atorvastatin inhibits human SV-SMC proliferation and invasion with IC50s of 0.39 μM and 2.39 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9576 Supplier Medchemexpress LLC Supplier No. HYB058950MG
HY-B0589 50mg Atorvastatin CAS: 134523-00-5 Atorvastatin is an orally active HMG-CoA reductase inhibitor, has the ability to effectively decrease blood lipids. Atorvastatin inhibits human SV-SMC proliferation and invasion with IC50s of 0.39 μM and 2.39 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.