Learn More
Selleck Chemical LLC Atorvastatin
SDP
Catalog No. 5000000033
Encompass_Preferred
Atorvastatin is a lipid lowering agent It is a competitive inhibitor of hydroxymethylglutaryl-coenzyme A (HMG-CoA) reductase the rate-determining enzyme in cholesterol biosynthesis via the mevalonate pathway Atorvastatin activates autophagy
Catalog No. 50-000-00033 Supplier Selleck Chemical LLC Supplier No. S57151G
Atorvastatin is a lipid lowering agent It is a competitive inhibitor of hydroxymethylglutaryl-coenzyme A (HMG-CoA) reductase the rate-determining enzyme in cholesterol biosynthesis via the mevalonate pathway Atorvastatin activates autophagy