Learn More
Selleck Chemical LLC Atorvastatin Calcium S2077-10mM/1mL
SDP
Catalog No. 503111132
Encompass_Preferred
Atorvastatin Calcium is an inhibitor of HMG-CoA reductase used as a cholesterol-lowering medication that blocks the production of cholesterol Atorvastatin Calcium induces apoptosis and autophagy
Catalog No. 50-311-1132 Supplier Selleck Chemical LLC Supplier No. S207710MM/1ML
Atorvastatin Calcium is an inhibitor of HMG-CoA reductase used as a cholesterol-lowering medication that blocks the production of cholesterol Atorvastatin Calcium induces apoptosis and autophagy