Learn More
Selleck Chemical LLC Atorvastatin Calcium S2077-500mg
SDP
Catalog No. 503111133
Encompass_Preferred
Atorvastatin Calcium is an inhibitor of HMG-CoA reductase used as a cholesterol-lowering medication that blocks the production of cholesterol Atorvastatin Calcium induces apoptosis and autophagy
Catalog No. 50-311-1133 Supplier Selleck Chemical LLC Supplier No. S2077500MG
Atorvastatin Calcium is an inhibitor of HMG-CoA reductase used as a cholesterol-lowering medication that blocks the production of cholesterol Atorvastatin Calcium induces apoptosis and autophagy