Learn More
Apexbio Technology LLC Atorvastatin Calcium, 500mg. Cas: 134523-03-8 MFCD: MFCD03613598. HMG-CoA reductase inhibitor
SDP
Supplier:  Apexbio Technology LLC A4367500
Encompass_Preferred
Atorvastatin Calcium is a potent inhibitor of HMG-CoA reductase with IC50 value of 150 nM[1].HMG-CoA reductase is the key enzyme of the mevalonate pathway which produces cholesterol. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4931