Learn More
Medchemexpress LLC Atorvastatin calcium hydrate | 344423-98-9 | 99.8% | 605.71 | 100 MG
SDP
Catalog No. 5000436650
Encompass_Preferred
Atorvastatin calcium salt trihydrate is a statin drug used to lower blood cholesterol levels. It works by inhibiting HMG-CoA reductase, an enzyme in the liver responsible for cholesterol production. It helps control high levels of low-density lipoprotein (LDL) cholesterol and triglycerides, thereby reducing the risk of heart attack and stroke. It is intended for research use only and not for sale to patients.
  • Belongs to the statin class of drugs.
  • Lowers blood cholesterol levels.
  • Has unique chemical properties for effective control of high LDL cholesterol and triglycerides.
  • Reduces the risk of heart attack and stroke.
  • Inhibits HMG-CoA reductase.
  • Biochemical reagent for life science-related research.
Catalog No. 50-004-36650 Supplier Medchemexpress LLC Supplier No. HY112490100MG
Atorvastatin calcium salt trihydrate is a statin drug used to lower blood cholesterol levels. It works by inhibiting HMG-CoA reductase, an enzyme in the liver responsible for cholesterol production. It helps control high levels of low-density lipoprotein (LDL) cholesterol and triglycerides, thereby reducing the risk of heart attack and stroke. It is intended for research use only and not for sale to patients.
  • Belongs to the statin class of drugs.
  • Lowers blood cholesterol levels.
  • Has unique chemical properties for effective control of high LDL cholesterol and triglycerides.
  • Reduces the risk of heart attack and stroke.
  • Inhibits HMG-CoA reductase.
  • Biochemical reagent for life science-related research.