Learn More
Medchemexpress LLC Atorvastatin calcium hydrate | 344423-98-9 | 99.8% | 605.71 | 250 MG
SDP
Catalog No. 5000439031
Encompass_Preferred
Atorvastatin calcium hydrate is a statin-class drug designed to lower blood cholesterol and triglyceride levels. It functions by inhibiting HMG-CoA reductase, an enzyme crucial for cholesterol production in the liver. This action helps reduce the risk of heart attack and stroke. It is intended for life science research applications.
  • Lowers blood cholesterol levels
  • Controls high levels of LDL cholesterol and triglycerides
  • Reduces risk of heart attack and stroke
  • Inhibits HMG-CoA reductase, an enzyme in the liver
  • Intended for life science research
Catalog No. 50-004-39031 Supplier Medchemexpress LLC Supplier No. 250MGHY112490
Atorvastatin calcium hydrate is a statin-class drug designed to lower blood cholesterol and triglyceride levels. It functions by inhibiting HMG-CoA reductase, an enzyme crucial for cholesterol production in the liver. This action helps reduce the risk of heart attack and stroke. It is intended for life science research applications.
  • Lowers blood cholesterol levels
  • Controls high levels of LDL cholesterol and triglycerides
  • Reduces risk of heart attack and stroke
  • Inhibits HMG-CoA reductase, an enzyme in the liver
  • Intended for life science research