Learn More
Medchemexpress LLC Atorvastatin calcium hydrate | 344423-98-9 | 99.83% | 1209.39 | 500 MG
SDP
Catalog No. 5000438255
Encompass_Preferred
Atorvastatin calcium salt trihydrate is a statin class drug used to lower blood cholesterol levels. It helps control high levels of low-density lipoprotein (LDL) cholesterol and triglycerides, thereby reducing the risk of heart attack and stroke. The drug works by inhibiting HMG-CoA reductase, an enzyme responsible for cholesterol production in the liver. It is also described as a biochemical reagent for life science related research.
  • Statin class drug
  • Lowers blood cholesterol levels
  • Controls high LDL cholesterol and triglycerides
  • Reduces risk of heart attack and stroke
  • Inhibits HMG-CoA reductase
  • Biochemical reagent for life science research
Catalog No. 50-004-38255 Supplier Medchemexpress LLC Supplier No. HY112490500MG
Atorvastatin calcium salt trihydrate is a statin class drug used to lower blood cholesterol levels. It helps control high levels of low-density lipoprotein (LDL) cholesterol and triglycerides, thereby reducing the risk of heart attack and stroke. The drug works by inhibiting HMG-CoA reductase, an enzyme responsible for cholesterol production in the liver. It is also described as a biochemical reagent for life science related research.
  • Statin class drug
  • Lowers blood cholesterol levels
  • Controls high LDL cholesterol and triglycerides
  • Reduces risk of heart attack and stroke
  • Inhibits HMG-CoA reductase
  • Biochemical reagent for life science research