Learn More
Medchemexpress LLC Atorvastatin calcium hydrate | 344423-98-9 | 99.83% | 50 MG
SDP
Catalog No. 5000437146
Encompass_Preferred
Atorvastatin calcium salt trihydrate is a statin-class drug used to lower blood cholesterol levels. It is effective in controlling high levels of low-density lipoprotein (LDL) cholesterol and triglycerides, which reduces the risk of heart attack and stroke. The drug works by inhibiting HMG-CoA reductase, an enzyme in the liver responsible for cholesterol production. It is intended for research use only.
  • Lowers blood cholesterol levels
  • Controls high levels of LDL cholesterol and triglycerides
  • Reduces risk of heart attack and stroke
  • Inhibits HMG-CoA reductase
  • Suitable for life science research
Catalog No. 50-004-37146 Supplier Medchemexpress LLC Supplier No. HY11249050MG
Atorvastatin calcium salt trihydrate is a statin-class drug used to lower blood cholesterol levels. It is effective in controlling high levels of low-density lipoprotein (LDL) cholesterol and triglycerides, which reduces the risk of heart attack and stroke. The drug works by inhibiting HMG-CoA reductase, an enzyme in the liver responsible for cholesterol production. It is intended for research use only.
  • Lowers blood cholesterol levels
  • Controls high levels of LDL cholesterol and triglycerides
  • Reduces risk of heart attack and stroke
  • Inhibits HMG-CoA reductase
  • Suitable for life science research