Learn More
Cayman Chemical Atorvastatin (calcium salt hydrate), Purity: ≥98%, CAS: 357164-38-6, 50 mg
SDP
Catalog No. 502889761
Encompass_Preferred
An HMG-CoA reductase inhibitor (IC50s = 73, 102, and 0.6 nM for HepG2 cells, human fibroblasts, and rat hepatocytes, respectively)
Catalog No. 50-288-9761 Supplier Cayman Chemical Supplier No. 1049350 MG
An HMG-CoA reductase inhibitor (IC50s = 73, 102, and 0.6 nM for HepG2 cells, human fibroblasts, and rat hepatocytes, respectively)