Learn More
Medchemexpress LLC Atorvastatin calcium salt trihydrate | 344423-98-9 | 99.8% | 605.71 | 1 G
SDP
Catalog No. 5000436340
Encompass_Preferred
Atorvastatin calcium salt trihydrate is a drug belonging to the statin class of drugs used to lower blood cholesterol levels. Atorvastatin calcium salt trihydrate has unique chemical properties that make it an effective tool in controlling high levels of low-density lipoprotein (LDL) cholesterol and triglycerides in the body, reducing the risk of heart attack and stroke. Atorvastatin calcium salt trihydrate works by inhibiting HMG-CoA reductase, the enzyme responsible for producing cholesterol in the liver.
  • Lowers blood cholesterol levels.
  • Controls high levels of low-density lipoprotein (LDL) cholesterol and triglycerides.
  • Reduces the risk of heart attack and stroke.
  • Works by inhibiting HMG-CoA reductase, the enzyme responsible for producing cholesterol in the liver.
  • Can be used as a biochemical reagent for life science-related research.
Catalog No. 50-004-36340 Supplier Medchemexpress LLC Supplier No. HY1124901G
Atorvastatin calcium salt trihydrate is a drug belonging to the statin class of drugs used to lower blood cholesterol levels. Atorvastatin calcium salt trihydrate has unique chemical properties that make it an effective tool in controlling high levels of low-density lipoprotein (LDL) cholesterol and triglycerides in the body, reducing the risk of heart attack and stroke. Atorvastatin calcium salt trihydrate works by inhibiting HMG-CoA reductase, the enzyme responsible for producing cholesterol in the liver.
  • Lowers blood cholesterol levels.
  • Controls high levels of low-density lipoprotein (LDL) cholesterol and triglycerides.
  • Reduces the risk of heart attack and stroke.
  • Works by inhibiting HMG-CoA reductase, the enzyme responsible for producing cholesterol in the liver.
  • Can be used as a biochemical reagent for life science-related research.