Learn More
Medchemexpress LLC Atorvastatin-d5 hemicalcium | 222412-82-0 | 99.0% | 500 UG
SDP
Supplier:  Medchemexpress LLC HYB0589S500UG
Encompass_Preferred
Atorvastatin-d5 hemicalcium is the deuterium labeled Atorvastatin. It is an orally active HMG-CoA reductase inhibitor that effectively decreases blood lipids. It also inhibits human SV-SMC proliferation and invasion.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Contains stable heavy isotopes for drug development quantitation
  • Deuteration can affect pharmacokinetic and metabolic profiles
  • Effectively decreases blood lipids
  • Inhibits human SV-SMC proliferation and invasion
Catalog No. 50-004-08300