Learn More
Medchemexpress LLC Atorvastatin-d5 (sodium) | 222412-87-5 | 96.95% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0589S11MG
Encompass_Preferred
Atorvastatin-d5 (sodium) is a deuterium-labeled Atorvastatin sodium. It acts as an orally active HMG-CoA reductase inhibitor, effectively decreasing blood lipids. This compound also inhibits human SV-SMC proliferation and invasion with IC50s of 0.39 μM and 2.39 μM respectively. Deuteration, the incorporation of stable heavy isotopes, can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Effectively decreases blood lipids
  • Inhibits human SV-SMC proliferation and invasion
Catalog No. 50-004-07759