Learn More
Medchemexpress LLC Atorvastatin epoxy tetrahydrofuran impurity | 873950-19-7 | 449.47 | 25 MG
SDP
Catalog No. 5000347840
Encompass_Preferred
Atorvastatin Epoxy Tetrahydrofuran Impurity is an impurity isolated oxidative degradation product of Atorvastatin. Atorvastatin is an orally active HMG-CoA reductase inhibitor, which has the ability to effectively decrease blood lipids. Atorvastatin treatment decreases apoptosis of myocardial cells by down-regulating GRP78, caspase-12 and CHOP expression in myocardial cells after myocardial infarction. The endoplasmic reticulum (ER) stress is activated in response to heart failure and angiotensin II (Ang II) stimulation.
  • Impurity isolated oxidative degradation product
  • Orally active HMG-CoA reductase inhibitor
  • Effectively decreases blood lipids
  • Decreases apoptosis of myocardial cells
  • Down-regulates GRP78, caspase-12 and CHOP expression
Catalog No. 50-003-47840 Supplier Medchemexpress LLC Supplier No. 25MGHY136185
Atorvastatin Epoxy Tetrahydrofuran Impurity is an impurity isolated oxidative degradation product of Atorvastatin. Atorvastatin is an orally active HMG-CoA reductase inhibitor, which has the ability to effectively decrease blood lipids. Atorvastatin treatment decreases apoptosis of myocardial cells by down-regulating GRP78, caspase-12 and CHOP expression in myocardial cells after myocardial infarction. The endoplasmic reticulum (ER) stress is activated in response to heart failure and angiotensin II (Ang II) stimulation.
  • Impurity isolated oxidative degradation product
  • Orally active HMG-CoA reductase inhibitor
  • Effectively decreases blood lipids
  • Decreases apoptosis of myocardial cells
  • Down-regulates GRP78, caspase-12 and CHOP expression