Learn More
Medchemexpress LLC Atorvastatin epoxy tetrahydrofuran impurity | 873950-19-7 | 449.47 | 50 MG
SDP
Catalog No. 5000350225
Encompass_Preferred
Atorvastatin epoxy tetrahydrofuran impurity is an impurity isolated from the oxidative degradation products of atorvastatin. Atorvastatin is an orally active HMG-CoA reductase inhibitor that effectively decreases blood lipids. This product is for research use only and not sold to patients.
  • Isolated from oxidative degradation products of atorvastatin.
  • Atorvastatin is an orally active HMG-CoA reductase inhibitor.
  • Effectively decreases blood lipids.
  • Reduces apoptosis of myocardial cells by down-regulating GRP78, caspase-12, and CHOP expression after myocardial infarction.
  • Involved in activation of endoplasmic reticulum (ER) stress in response to heart failure and angiotensin II (Ang II) stimulation.
Catalog No. 50-003-50225 Supplier Medchemexpress LLC Supplier No. HY13618550MG
Atorvastatin epoxy tetrahydrofuran impurity is an impurity isolated from the oxidative degradation products of atorvastatin. Atorvastatin is an orally active HMG-CoA reductase inhibitor that effectively decreases blood lipids. This product is for research use only and not sold to patients.
  • Isolated from oxidative degradation products of atorvastatin.
  • Atorvastatin is an orally active HMG-CoA reductase inhibitor.
  • Effectively decreases blood lipids.
  • Reduces apoptosis of myocardial cells by down-regulating GRP78, caspase-12, and CHOP expression after myocardial infarction.
  • Involved in activation of endoplasmic reticulum (ER) stress in response to heart failure and angiotensin II (Ang II) stimulation.