Learn More
Medchemexpress LLC Atorvastatin epoxy tetrahydrofuran impurity | 873950-19-7 | MFCD31700699 | 96.0% | C26H24FNO5 | 10MG
SDP
Catalog No. 5000258797
Encompass_Preferred
Atorvastatin epoxy tetrahydrofuran impurity is an oxidative degradation product of atorvastatin provided as an analytical reference standard for research use. Supplied in milligram quantities, it supports impurity profiling, method development, and stability testing. Users should consult the lot-specific Certificate of Analysis for purity and storage conditions.
  • Used as an impurity reference for analytical method development.
  • Suitable for stability studies and degradation profiling.
  • Provided in small, laboratory-scale quantities for research use.
  • Chemical formula C26H24FNO5 and CAS 873950-19-7 for unambiguous identification.
  • Lot-specific purity and storage conditions available in the Certificate of Analysis.
Catalog No. 50-002-58797 Supplier Medchemexpress LLC Supplier No. HY13618510MG
Atorvastatin epoxy tetrahydrofuran impurity is an oxidative degradation product of atorvastatin provided as an analytical reference standard for research use. Supplied in milligram quantities, it supports impurity profiling, method development, and stability testing. Users should consult the lot-specific Certificate of Analysis for purity and storage conditions.
  • Used as an impurity reference for analytical method development.
  • Suitable for stability studies and degradation profiling.
  • Provided in small, laboratory-scale quantities for research use.
  • Chemical formula C26H24FNO5 and CAS 873950-19-7 for unambiguous identification.
  • Lot-specific purity and storage conditions available in the Certificate of Analysis.