Learn More
Medchemexpress LLC Atorvastatin ethyl ester | 1146977-93-6 | MFCD28143549 | 98.2% | 586.69 g/mol | C35H39FN2O5 | 5 MG
SDP
Catalog No. 5000223344
Encompass_Preferred
Atorvastatin ethyl ester is a research chemical derivative of atorvastatin reported to strongly inhibit 9-cis-retinoic acid (9-cis-RA)-induced Gal4 reporter activity. It is intended for preclinical research use and is characterized by CAS 1146977-93-6, molecular formula C35H39FN2O5, and molecular weight 586.69 g/mol.
  • Reported inhibitor of 9-cis-RA-induced Gal4 reporter activity.
  • Intended for research use only, not for human consumption.
  • High purity (98.2%).
  • Characterized by CAS 1146977-93-6 and formula C35H39FN2O5.
  • Available in small pack sizes suitable for assay development, including five mg packs.
Catalog No. 50-002-23344 Supplier Medchemexpress LLC Supplier No. HY1353785MG
Atorvastatin ethyl ester is a research chemical derivative of atorvastatin reported to strongly inhibit 9-cis-retinoic acid (9-cis-RA)-induced Gal4 reporter activity. It is intended for preclinical research use and is characterized by CAS 1146977-93-6, molecular formula C35H39FN2O5, and molecular weight 586.69 g/mol.
  • Reported inhibitor of 9-cis-RA-induced Gal4 reporter activity.
  • Intended for research use only, not for human consumption.
  • High purity (98.2%).
  • Characterized by CAS 1146977-93-6 and formula C35H39FN2O5.
  • Available in small pack sizes suitable for assay development, including five mg packs.