Learn More
Medchemexpress LLC Atorvastatin ethyl ester | 1146977-93-6 | MFCD28143549 | 98.2% | C35H39FN2O5 | 10MG
SDP
Catalog No. 5000261310
Encompass_Preferred
Atorvastatin ethyl ester is an ethyl-ester derivative of atorvastatin supplied for research use. It strongly inhibits 9-cis-retinoic acid-induced Gal4 reporter activity and is used as a biochemical probe and analytical reference. The compound has molecular formula C35H39FN2O5, CAS 1146977-93-6, and a molecular weight of 586.69 g·mol⁻¹.
  • Inhibits 9-cis-RA-induced Gal4 reporter activity.
  • Provided in multiple small-scale pack sizes for laboratory research.
  • High reported purity (98.16%).
  • Molecular formula C35H39FN2O5 and molecular weight 586.69 g·mol⁻¹.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-61310 Supplier Medchemexpress LLC Supplier No. HY13537810MG
Atorvastatin ethyl ester is an ethyl-ester derivative of atorvastatin supplied for research use. It strongly inhibits 9-cis-retinoic acid-induced Gal4 reporter activity and is used as a biochemical probe and analytical reference. The compound has molecular formula C35H39FN2O5, CAS 1146977-93-6, and a molecular weight of 586.69 g·mol⁻¹.
  • Inhibits 9-cis-RA-induced Gal4 reporter activity.
  • Provided in multiple small-scale pack sizes for laboratory research.
  • High reported purity (98.16%).
  • Molecular formula C35H39FN2O5 and molecular weight 586.69 g·mol⁻¹.
  • Intended for research use only; not for human or veterinary use.