Learn More
Medchemexpress LLC HY-17379 50mg , Atorvastatin (hemicalcium salt) CAS:134523-03-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1737950MG
Encompass_Preferred
HY-17379 50mg Atorvastatin (hemicalcium salt) CAS: 134523-03-8 Atorvastatin hemicalcium salt (CI-981) is an orally active 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase inhibitor, has the ability to effectively decrease blood lipids. Atorvastatin hemicalcium salt inhibits human SV-SMC proliferation and invasion with IC50s of 0.39 μM and 2.39 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9553