Learn More
Medchemexpress LLC Atorvastatin hemicalcium | 134523-03-8 | 99.94% | 577.67 | 1 ML
SDP
Catalog No. 5000384573
Encompass_Preferred
Atorvastatin hemicalcium salt (CI-981) is an orally active 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase inhibitor. It is capable of penetrating the blood-brain barrier and effectively reduces blood lipids. This compound inhibits human SV-SMC proliferation and invasion, and decreases apoptosis of myocardial cells by down-regulating specific proteins. It also activates endoplasmic reticulum (ER) stress in response to heart failure and angiotensin II (Ang II) stimulation. This product is for research use only.
  • Orally active HMG-CoA reductase inhibitor
  • Penetrates the blood-brain barrier
  • Effectively reduces blood lipids
  • Inhibits human SV-SMC proliferation and invasion
  • Decreases myocardial cell apoptosis
  • Activates endoplasmic reticulum (ER) stress response
  • Reduces ER stress signaling proteins
  • Inhibits proinflammatory cytokines
Catalog No. 50-003-84573 Supplier Medchemexpress LLC Supplier No. HY1737910MM/1ML
Atorvastatin hemicalcium salt (CI-981) is an orally active 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase inhibitor. It is capable of penetrating the blood-brain barrier and effectively reduces blood lipids. This compound inhibits human SV-SMC proliferation and invasion, and decreases apoptosis of myocardial cells by down-regulating specific proteins. It also activates endoplasmic reticulum (ER) stress in response to heart failure and angiotensin II (Ang II) stimulation. This product is for research use only.
  • Orally active HMG-CoA reductase inhibitor
  • Penetrates the blood-brain barrier
  • Effectively reduces blood lipids
  • Inhibits human SV-SMC proliferation and invasion
  • Decreases myocardial cell apoptosis
  • Activates endoplasmic reticulum (ER) stress response
  • Reduces ER stress signaling proteins
  • Inhibits proinflammatory cytokines