Learn More
Medchemexpress LLC Atorvastatin hemicalcium salt | 134523-03-8 | 99.9% | 577.67 | 200 MG
SDP
Catalog No. 5000384720
Encompass_Preferred
Atorvastatin hemicalcium salt (CI-981) is an orally active 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase inhibitor, with blood-brain barrier permeability. It has the ability to effectively decrease blood lipids. This compound inhibits human SV-SMC proliferation and invasion with IC50s of 0.39 μM and 2.39 μM, respectively.
  • Orally active 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase inhibitor.
  • Possesses blood-brain barrier permeability.
  • Effectively decreases blood lipids.
  • Inhibits human SV-SMC proliferation and invasion.
Catalog No. 50-003-84720 Supplier Medchemexpress LLC Supplier No. HY17379200MG
Atorvastatin hemicalcium salt (CI-981) is an orally active 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase inhibitor, with blood-brain barrier permeability. It has the ability to effectively decrease blood lipids. This compound inhibits human SV-SMC proliferation and invasion with IC50s of 0.39 μM and 2.39 μM, respectively.
  • Orally active 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase inhibitor.
  • Possesses blood-brain barrier permeability.
  • Effectively decreases blood lipids.
  • Inhibits human SV-SMC proliferation and invasion.