Learn More
Medchemexpress LLC Atorvastatin (hemicalcium salt) | 134523-03-8 | 99.9% | 577.67 | 500 MG
SDP
Catalog No. 5000384778
Encompass_Preferred
Atorvastatin hemicalcium salt is an orally active 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase inhibitor with blood-brain barrier permeability. It effectively decreases blood lipids and inhibits human SV-SMC proliferation and invasion. This compound reduces apoptosis of myocardial cells by down-regulating GRP78, caspase-12, and CHOP expression, and activates endoplasmic reticulum stress in response to heart failure and angiotensin II stimulation.
  • Orally active HMG-CoA reductase inhibitor
  • Blood-brain barrier permeable
  • Decreases blood lipids
  • Inhibits SV-SMC proliferation and invasion
  • Reduces apoptosis of myocardial cells
  • Activates endoplasmic reticulum stress response
  • Inhibits proinflammatory cytokines
Catalog No. 50-003-84778 Supplier Medchemexpress LLC Supplier No. HY17379500MG
Atorvastatin hemicalcium salt is an orally active 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase inhibitor with blood-brain barrier permeability. It effectively decreases blood lipids and inhibits human SV-SMC proliferation and invasion. This compound reduces apoptosis of myocardial cells by down-regulating GRP78, caspase-12, and CHOP expression, and activates endoplasmic reticulum stress in response to heart failure and angiotensin II stimulation.
  • Orally active HMG-CoA reductase inhibitor
  • Blood-brain barrier permeable
  • Decreases blood lipids
  • Inhibits SV-SMC proliferation and invasion
  • Reduces apoptosis of myocardial cells
  • Activates endoplasmic reticulum stress response
  • Inhibits proinflammatory cytokines