Learn More
Abcam Atorvastatin hemicalcium salt, HMG-CoA reductase inhibitor, 10MG
Catalog No. 502706924
Encompass_Preferred
MW 1155.3 Da, Purity >99%. Potent HMG-CoA reductase inhibitor (IC₅₀ = 8 nM). Metabolised by CYP3A4. Able to reduce circulating LDL-C through inhibition of cholesterol biosynthesis and inducing expression of LDL receptors.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6924 Supplier Abcam Supplier No. AB14108310MG
MW 1155.3 Da, Purity >99%. Potent HMG-CoA reductase inhibitor (IC₅₀ = 8 nM). Metabolised by CYP3A4. Able to reduce circulating LDL-C through inhibition of cholesterol biosynthesis and inducing expression of LDL receptors.
The product is subject to the following: Abcam Restricted Use Statement