Learn More
Medchemexpress LLC Atorvastatin (hemicalcium trihydrate) | 344920-08-7 | 100 MG
SDP
Catalog No. 5000405909
Encompass_Preferred
Atorvastatin hemicalcium trihydrate is an orally active inhibitor of HMG-CoA reductase. Its primary function is to effectively lower blood lipid levels. Additionally, it has been observed to inhibit human SV-SMC proliferation and invasion, with IC50 values of 0.39 μM and 2.39 μM, respectively.
  • Orally active HMG-CoA reductase inhibitor
  • Effectively decreases blood lipids
  • Inhibits human SV-SMC proliferation and invasion
  • Demonstrates IC50s of 0.39 μM (proliferation) and 2.39 μM (invasion) in human SV-SMC
Catalog No. 50-004-05909 Supplier Medchemexpress LLC Supplier No. HYB0589E100MG
Atorvastatin hemicalcium trihydrate is an orally active inhibitor of HMG-CoA reductase. Its primary function is to effectively lower blood lipid levels. Additionally, it has been observed to inhibit human SV-SMC proliferation and invasion, with IC50 values of 0.39 μM and 2.39 μM, respectively.
  • Orally active HMG-CoA reductase inhibitor
  • Effectively decreases blood lipids
  • Inhibits human SV-SMC proliferation and invasion
  • Demonstrates IC50s of 0.39 μM (proliferation) and 2.39 μM (invasion) in human SV-SMC