Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Atorvastatin lactone | 125995-03-1 | MFCD00899262 | 96.7% | 540.62 | C33H33FN2O4 | 1 ML
SDP

Supplier:  Medchemexpress LLC HY10187310MM 1ML

Encompass_Preferred

Atorvastatin lactone is the lactone (prodrug) form of atorvastatin and a small-molecule HMG-CoA reductase inhibitor supplied as a ready-to-use analytical/research reagent. This product is offered as a prepared solution for biochemical assays and analytical workflows, with accompanying certificate of analysis.

  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • Verified purity 96.7% by HPLC.
  • Molecular weight 540.62 g·mol-1 and formula C33H33FN2O4.
  • Supplied with certificate of analysis.
  • Storage: -80°C for up to 6 months, -20°C for up to 1 month.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-45562


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.