Learn More
Medchemexpress LLC Atorvastatin lactone | 125995-03-1 | MFCD00899262 | 96.7% | 540.62 g·mol-1 | C33H33FN2O4 | 5 MG
SDP
Catalog No. 5000248494
Encompass_Preferred
Atorvastatin lactone is the lactone (prodrug) form of atorvastatin, acting as an HMG-CoA reductase inhibitor used in research on cholesterol biosynthesis, statin metabolism, and enzymatic assays. Supplied as a purified solid, it is typically prepared as a DMSO stock for in vitro and analytical applications.
  • Prodrug/lactone form of atorvastatin for metabolic and pharmacology studies.
  • High purity suitable for analytical and biochemical assays.
  • Powder form; soluble in DMSO for stock preparations.
  • Molecular weight approximately 540.62 g·mol-1.
  • Used as an HMG-CoA reductase inhibitor in research applications.
Catalog No. 50-002-48494 Supplier Medchemexpress LLC Supplier No. HY1018735MG
Atorvastatin lactone is the lactone (prodrug) form of atorvastatin, acting as an HMG-CoA reductase inhibitor used in research on cholesterol biosynthesis, statin metabolism, and enzymatic assays. Supplied as a purified solid, it is typically prepared as a DMSO stock for in vitro and analytical applications.
  • Prodrug/lactone form of atorvastatin for metabolic and pharmacology studies.
  • High purity suitable for analytical and biochemical assays.
  • Powder form; soluble in DMSO for stock preparations.
  • Molecular weight approximately 540.62 g·mol-1.
  • Used as an HMG-CoA reductase inhibitor in research applications.