Learn More
TARGETMOL CHEMICALS INC Atorvastatin lactone 25MG
SDP
Catalog No. 502880499
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Atorvastatin lactone is a prodrug form of atorvastatin. Which is an orally active 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase inhibitor. Purity 98.16%
Catalog No. 50-288-0499 Supplier TARGETMOL CHEMICALS INC Supplier No. T1434225MG
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Atorvastatin lactone is a prodrug form of atorvastatin. Which is an orally active 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase inhibitor. Purity 98.16%