Learn More
Medchemexpress LLC Atorvastatin methyl ester | 345891-62-5 | MFCD21363420 | 98.8% | C34H37FN2O5 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13537610MG
Encompass_Preferred
Atorvastatin methyl ester is a methyl-esterified derivative of atorvastatin provided for laboratory research. It is used as a drug derivative and has been reported to inhibit 9-cis-RA-induced Gal4 reporter activity more strongly than atorvastatin. The compound is supplied as solids and as a DMSO solution for in vitro experiments.
  • Methyl-esterified derivative of atorvastatin used for research
  • Reported to inhibit 9-cis-RA-induced Gal4 reporter activity more strongly than atorvastatin
  • Available as solid quantities (10 mg-500 mg) and 10 mM solution in DMSO
  • High purity (approx. 98.8%) suitable for in vitro studies
  • Useful structural analogue for metabolism and activity studies
Catalog No. 50-002-60037