Learn More
Medchemexpress LLC Atorvastatin-PEG3-FITC | 1440755-31-6 | 98.2% | C64H68FN5O12S | 10MG
SDP
Catalog No. 5000261802
Encompass_Preferred
Atorvastatin-PEG3-FITC is a fluorescently labeled derivative of atorvastatin used as a ligand in fluorescence anisotropy assays and reported as a KRAS-PDEδ interaction inhibitor (compound S31). It is provided for research use with defined chemical characteristics and high reported purity suitable for biochemical experiments.
  • Acts as a ligand in fluorescence anisotropy assays.
  • Reported to inhibit KRAS-PDEδ interaction (compound S31).
  • Contains an FITC fluorescent tag for fluorescence-based detection.
  • High reported purity (98.17%) suitable for biochemical assays.
  • Defined molecular formula and molecular weight for assay calculations.
  • Available in small research pack sizes for assay optimization.
Catalog No. 50-002-61802 Supplier Medchemexpress LLC Supplier No. HY13497710MG
Atorvastatin-PEG3-FITC is a fluorescently labeled derivative of atorvastatin used as a ligand in fluorescence anisotropy assays and reported as a KRAS-PDEδ interaction inhibitor (compound S31). It is provided for research use with defined chemical characteristics and high reported purity suitable for biochemical experiments.
  • Acts as a ligand in fluorescence anisotropy assays.
  • Reported to inhibit KRAS-PDEδ interaction (compound S31).
  • Contains an FITC fluorescent tag for fluorescence-based detection.
  • High reported purity (98.17%) suitable for biochemical assays.
  • Defined molecular formula and molecular weight for assay calculations.
  • Available in small research pack sizes for assay optimization.