Learn More
Medchemexpress LLC Atorvastatin sodium | 134523-01-6 | MFCD08458204 | 99.9% | 580.6 g/mol | C33H34FN2NaO5 | 1 ML
SDP
Catalog No. 5000242341
Encompass_Preferred
Atorvastatin sodium is the sodium salt of atorvastatin, an HMG-CoA reductase inhibitor used in research on lipid metabolism, cardiovascular biology, and inflammation. It is supplied at high purity and is available as solids and as a 10 mM solution in DMSO for biochemical and cell-based assays.
  • High purity suitable for research (≈99.9%).
  • Available as 10 mM DMSO solution for ready-to-use dosing.
  • Also available as multiple solid weights for formulation or storage.
  • Used in assays targeting HMG-CoA reductase and related pathways.
  • Documented activity in cellular and in vivo cardiovascular models.
Catalog No. 50-002-42341 Supplier Medchemexpress LLC Supplier No. HY10825710MM 1ML
Atorvastatin sodium is the sodium salt of atorvastatin, an HMG-CoA reductase inhibitor used in research on lipid metabolism, cardiovascular biology, and inflammation. It is supplied at high purity and is available as solids and as a 10 mM solution in DMSO for biochemical and cell-based assays.
  • High purity suitable for research (≈99.9%).
  • Available as 10 mM DMSO solution for ready-to-use dosing.
  • Also available as multiple solid weights for formulation or storage.
  • Used in assays targeting HMG-CoA reductase and related pathways.
  • Documented activity in cellular and in vivo cardiovascular models.