Learn More
Medchemexpress LLC Atorvastatin sodium | 134523-01-6 | MFCD08458204 | 99.9% | 580.62 | C33H34FN2NaO5 | 10 MG
SDP
Catalog No. 5000244072
Encompass_Preferred
Atorvastatin sodium is the sodium salt form of atorvastatin, an HMG-CoA reductase inhibitor used in research to study cholesterol biosynthesis, lipid metabolism, and related cellular effects. It is supplied as a high-purity white to off-white solid and is characterized by standard analytical data including HPLC and molecular weight determinations.
  • High purity (HPLC) for reproducible assay results.
  • Molecular formula C33H34FN2NaO5 and molecular weight 580.62.
  • White to off-white solid appearance suitable for handling and storage.
  • Suitable for biochemical and cellular studies of HMG-CoA reductase activity.
  • Batch-specific certificate of analysis available for quality verification.
Catalog No. 50-002-44072 Supplier Medchemexpress LLC Supplier No. HY10825710MG
Atorvastatin sodium is the sodium salt form of atorvastatin, an HMG-CoA reductase inhibitor used in research to study cholesterol biosynthesis, lipid metabolism, and related cellular effects. It is supplied as a high-purity white to off-white solid and is characterized by standard analytical data including HPLC and molecular weight determinations.
  • High purity (HPLC) for reproducible assay results.
  • Molecular formula C33H34FN2NaO5 and molecular weight 580.62.
  • White to off-white solid appearance suitable for handling and storage.
  • Suitable for biochemical and cellular studies of HMG-CoA reductase activity.
  • Batch-specific certificate of analysis available for quality verification.