Learn More
TARGETMOL CHEMICALS INC ATORVASTATIN SODIUM 50MG
SDP
Catalog No. 502577230
Encompass_Preferred
Also available in 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Atorvastatin Sodium (Lipitor) is a competitive inhibitor of HMG-CoA reductase and increases the expression of low density lipoprotein (LDL) receptors on hepatocytes. Atorvastatin Sodium treatment inhibits aquaporin 4 to reduce ischaemic brain oedema. purity: 99%
Catalog No. 50-257-7230 Supplier TARGETMOL CHEMICALS INC Supplier No. T2088750MG
Also available in 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Atorvastatin Sodium (Lipitor) is a competitive inhibitor of HMG-CoA reductase and increases the expression of low density lipoprotein (LDL) receptors on hepatocytes. Atorvastatin Sodium treatment inhibits aquaporin 4 to reduce ischaemic brain oedema. purity: 99%