Learn More
Medchemexpress LLC HY-17572A 5mg Medchemexpress, Atosiban (acetate) CAS:914453-95-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY17572A5MG
Encompass_Preferred
Medchemexpress, HY-17572A 5mg Atosiban (acetate) CAS:914453-95-5 Atosiban acetate (RW22164 acetate;RWJ22164 acetate) is a nonapeptide competitive vasopressin/oxytocin receptor antagonist, and is a desamino-oxytocin analogue. Atosiban is the main tocolytic agent and has the potential for spontaneous preterm labor research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1990