Learn More
Medchemexpress LLC Atosiban (acetate) | 914453-95-5 | 99.8% | 1054.24 | 50 MG
SDP
Catalog No. 5000366163
Encompass_Preferred
Atosiban acetate is a nonapeptide competitive vasopressin/oxytocin receptor antagonist and a desamino-oxytocin analogue. It is the main tocolytic agent and has the potential for research into spontaneous preterm labor.
  • Competitive vasopressin/oxytocin receptor antagonist and desamino-oxytocin analogue
  • Functions as the main tocolytic agent
  • Suitable for research into spontaneous preterm labor
  • Inhibits oxytocin-mediated IP3 release from myometrial cells
  • Reduces intracellular calcium release and influx
  • Suppresses oxytocin-mediated prostaglandin release
  • Influences feto-maternal cardiovascular and renal systems
  • Blocks oxytocin-receptor-expressing neuron activation
Catalog No. 50-003-66163 Supplier Medchemexpress LLC Supplier No. 50MGHY17572A
Atosiban acetate is a nonapeptide competitive vasopressin/oxytocin receptor antagonist and a desamino-oxytocin analogue. It is the main tocolytic agent and has the potential for research into spontaneous preterm labor.
  • Competitive vasopressin/oxytocin receptor antagonist and desamino-oxytocin analogue
  • Functions as the main tocolytic agent
  • Suitable for research into spontaneous preterm labor
  • Inhibits oxytocin-mediated IP3 release from myometrial cells
  • Reduces intracellular calcium release and influx
  • Suppresses oxytocin-mediated prostaglandin release
  • Influences feto-maternal cardiovascular and renal systems
  • Blocks oxytocin-receptor-expressing neuron activation