Learn More
Cayman Chemical Atovaquone-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 2070015-14-2, 1 mg
SDP
Catalog No. 502938451
Encompass_Preferred
An internal standard for the quantification of atovaquone by GC- or LC-MS
Catalog No. 50-293-8451 Supplier Cayman Chemical Supplier No. 284911 MG
An internal standard for the quantification of atovaquone by GC- or LC-MS