Learn More
Apexbio Technology LLC Atovaquone(Synonyms: Mepron, Wellvone, 566C80, BW 566C, Atavaquone, Hydroxy-1,4-naphthoquinone derivative), 50mg, CAS: 95233-18-4.
SDP
Catalog No. 5000060412
Encompass_Preferred
Atovaquone (CAS 95233-18-4) is a hydroxynaphthoquinone compound employed in biomedical research primarily due to its inhibitory action against mitochondrial electron transport in pathogenic microorganisms Specifically it targets cytochrome bc1 complex (complex III) disrupting mitochondrial respiration and consequently impairing pyrimidine biosynthesis and replication in parasites This mechanism underlies its research utility in studying parasite metabolism and antiprotozoal therapeutics notably concerning Pneumocystis jirovecii pneumonia toxoplasmosis malaria and babesiosis
Catalog No. 50-000-60412 Supplier Apexbio Technology LLC Supplier No. B207850
Atovaquone (CAS 95233-18-4) is a hydroxynaphthoquinone compound employed in biomedical research primarily due to its inhibitory action against mitochondrial electron transport in pathogenic microorganisms Specifically it targets cytochrome bc1 complex (complex III) disrupting mitochondrial respiration and consequently impairing pyrimidine biosynthesis and replication in parasites This mechanism underlies its research utility in studying parasite metabolism and antiprotozoal therapeutics notably concerning Pneumocystis jirovecii pneumonia toxoplasmosis malaria and babesiosis