Learn More
TARGETMOL CHEMICALS INC Atovaquone 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T149150MG
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Atovaquone (Atavaquone) is a hydroxynaphthoquinone that has antimicrobial activity and is being used in antimalarial protocols. Purity 99.53%
Catalog No. 50-287-6261