Learn More
Medchemexpress LLC Atovaquone | 95233-18-4 | 99.75% | 366.84 | 50 MG
SDP
Catalog No. 5000349444
Encompass_Preferred
Atovaquone is a potent, selective, and orally active inhibitor targeting the parasite's mitochondrial cytochrome bc1 complex. It is effective against human and *P. falciparum* cytochrome bc1 activity and is recognized as an antimalarial agent.
  • Potent, selective, and orally active inhibitor of the parasite's mitochondrial cytochrome bc1 complex
  • Effective against human and *P. falciparum* cytochrome bc1 activity
  • Functions as an antimalarial agent
  • Potential for investigation of pneumocystis pneumonia, toxoplasmosis, malaria, and babesia
  • Targets the Qo site of the *Plasmodium* cytochrome bc1 complex within the mitochondrial electron transport chain
  • Demonstrates transmission blocking potential against mosquito development
Catalog No. 50-003-49444 Supplier Medchemexpress LLC Supplier No. HY1383250MG
Atovaquone is a potent, selective, and orally active inhibitor targeting the parasite's mitochondrial cytochrome bc1 complex. It is effective against human and *P. falciparum* cytochrome bc1 activity and is recognized as an antimalarial agent.
  • Potent, selective, and orally active inhibitor of the parasite's mitochondrial cytochrome bc1 complex
  • Effective against human and *P. falciparum* cytochrome bc1 activity
  • Functions as an antimalarial agent
  • Potential for investigation of pneumocystis pneumonia, toxoplasmosis, malaria, and babesia
  • Targets the Qo site of the *Plasmodium* cytochrome bc1 complex within the mitochondrial electron transport chain
  • Demonstrates transmission blocking potential against mosquito development